Real-Time PCR kit for detection of Vibrio vulnificus
Leverancier: Q BIOANALYTIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
QB-RTI-09-1000 QBRTI-09-10SMP QB-RTI-09-250 QB-RTI-09-50
710-1741EA 0 EUR
710-1741 710-1742 710-1743 710-1744
Real-Time PCR kit for detection of Vibrio vulnificus
Testsystemen voor microbiologie PCR testen
Vibrio vulnificus causes severe wound infections and is harmful when ingested. 33% of patients being infected die. The detection in food and water samples using this Real-Time PCR Test is very rapid and versatile. You have results available already after 24 hours. You can use this test in combination with an appropriate pre- enrichment in alkaline peptone-water or you can confirm bacterial colonies on agar plates.
  • TaqMan probes
  • Contains internal amplification control in HEX channel
  • Taget in FAM channel
  • Contains UNG to protect from re-amplification
  • Positive DNA and negative control included
Certificaten: DIN EN ISO 20838 conform detection in food, feed and water samples from pre-enrichment cultures or for confirmation of bacterial colonies