Real-Time PCR kit for detection of Pseudomonas aeruginosa
Leverancier: Q BIOANALYTIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
QB-RT-23-1000 QB-RT-23-250
QBIAQB-RT-23-1000EA 0 EUR
QBIAQB-RT-23-1000 QBIAQB-RT-23-250
Real-Time PCR kit for detection of Pseudomonas aeruginosa
Testsystemen voor microbiologie PCR testen
Pseudomonas aeruginosa is tested usually with cultural methods. However, this may take several days to deliver results. Using this Real-Time PCR test you may have results already after a few hours using a DNA purification directly from filtered water without pre-enrichment. You can also use this test in combination with an appropriate pre- enrichment according to DIN EN ISO 20837. The kit is also suitable to confirm bacterial colonies on agar plates.
  • TaqMan probes
  • Taget in FAM channel
  • Contains UNG to protect from re-amplification
  • Positive DNA and negative control included
Certificaten: DIN EN ISO 20838 conform detection in food and feed samples from pre-enrichment cultures or for confirmation of bacterial colonies.