Real-Time PCR kit for screening for genetically modified organisms in food and feed
Leverancier: Q BIOANALYTIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
QB-RT-91-10 QB-RT-91-50 QB-RT-91-1000
QBIAQB-RT-91-10EA 0 EUR
QBIAQB-RT-91-10 710-1720 707-0043
Real-Time PCR kit for screening for genetically modified organisms in food and feed
Testsystemen voor microbiologie PCR testen
The Real-Time PCR screening kit is addressing three different targets: 35S promoter; NOS Terminator; CTP2-CP4 EPSPS. The tests are all in accordance with the JRC guideline. Advantage of the CTP2-CP4 EPSPS compared to FMV detection is that the used target includes artificial sequences, which are not present naturally like the virus sequence of FMV.
  • TaqMan probes
  • Taget in FAM channel
  • Contains UNG to protect from re-amplification
  • Positive DNA and negative control included
Certificaten: GMO Screening according to JRC guidelines.