Real-Time PCR kit for detection of Staphylococcus aureus
Leverancier: Q BIOANALYTIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
QB-RTI-04-10 QB-RTI-04-1000 QB-RTI-04-250 QB-RTI-04-50
710-1737EA 0 EUR
710-1737 710-1738 710-1739 710-1740
Real-Time PCR kit for detection of Staphylococcus aureus
Testsystemen voor microbiologie PCR testen
Real-Time kit for the detection of Staphylococcus aureus.
  • TaqMan probes
  • Contains internal amplification control in HEX channel
  • Taget in FAM channel
  • Contains UNG to protect from re-amplification
  • Positive DNA and negative control included
Certificaten: DIN EN ISO 20838 conform detection in food and feed samples from pre-enrichment cultures or for confirmation of bacterial colonies.
Order Now
Specificaties
Environmentally Preferable