Dehydrated culture media, L-Broth, Molecular Biology Certified™, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
113001031 113001021
ICNA113001031EA 298 EUR
ICNA113001031 ICNA113001021
Dehydrated culture media, L-Broth, Molecular Biology Certified™, MP Biomedicals
Media voor microbiologie Gedehydrateerde media voor microbiologie
L-Broth Medium is a nutritionally rich medium originally developed for growth and maintenance of E. coli strains.
  • Available as capsules or powder
  • Molecular biology certified
L-Broth Medium supports a luxurious growth of E. coli cells as tryptone and yeast extract supply essential growth factors that E. coli cells would otherwise have to synthesise. The medium also contains essential electrolytes for transport and osmotic balance, due to the NaCl component.