Dehydrated culture media, EMM media, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
114110712 114110022 114110032 114110012
ICNA114110712EA 194 EUR
ICNA114110712 ICNA114110022 ICNA114110032 ICNA114110012
Dehydrated culture media, EMM media, MP Biomedicals
Media voor microbiologie Gedehydrateerde media voor microbiologie
EMM (Edinburgh Minimal Medium) is a minimal defined medium for the growth of fission yeast Schizosaccharomyces pombe.