Dehydrated culture media, CSM media, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
114520722 114511222 114511212 114500012
ICNA114520722EA 0 EUR
ICNA114520722 ICNA114511222 ICNA114511212 ICNA114500012
Dehydrated culture media, CSM media, MP Biomedicals
Media voor microbiologie Gedehydrateerde media voor microbiologie
Complete Supplement Mixture (CSM) is a culture growth media supplement for virtually all strains of Saccharomyces cerevisiae.
  • Available as single, double, triple, four, four+ dropout formulations of aminoacids
  • Allows selection for auxotrophic requirements and transformants
CSM is time-tested dropout supplement for growth of S. cerevisiae. Cells grow vigorously in CSM supplemented medium which is easily made by adding the CSM powder to DOB of DOBA base medium and autoclaving.
This is used to select for yeast containing plasmids with specific nutritional markers and to select for gene function in transformation experiments.