Thrombin (biotinylated)
Leverancier: Merck Millipore (Novagen)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
69672-3 69672-1000UN 69672-500UN
69672-3EA 427 EUR
69672-3 69672-1000UN 69672-500UN
Thrombin (biotinylated)
Enzymen
Biotinylated Thrombin has covalently attached biotin for easy removal of the enzyme from cleavage reactions using immobilised streptavidin. This preparation is tested for activity using the same assay as for unmodified thrombin, and for >99% binding to streptavidin agarose.
Storage temperature: −20 °C.