Extraction reagents
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ADI801502 ADI801526
ENZOADI801502EA 171 EUR
ENZOADI801502 ENZOADI801526
Extraction reagents
Buffers Extractiebuffers
Concentrated lysis buffer for preparation of cell and tissue extracts.
TRIS buffer containing detergents.