Potato dextrose broth, for microbiology
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P6685-1KG P6685-250G
SIALP6685-1KGEA 576 EUR
SIALP6685-1KG SIALP6685-250G
Potato dextrose broth, for microbiology
Media voor microbiologie Gedehydrateerde media voor microbiologie
Contains infusion from potatoes plus glucose.
  • pH (24 g/l, water): 4,2 –6,2
Recommended use at 24,0 g per litre.
Verpakking: Packed in poly bottle.
Order Now
Specificaties
Environmentally Preferable