Brefeldin A ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

B3820-10MG B3820-50MG
TCIAB3820-10MGEA 131 EUR
TCIAB3820-10MG TCIAB3820-50MG
Brefeldin A ≥98.0% (by HPLC)
Brefeldin A
Formule: C₁₆H₂₄O₄
Smeltpunt: 204°C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00083258
CAS nummer: 20350-15-6
UN: 2811
ADR: 6.1,III
Merck Index: 14,01369

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 98.0 area%
Elemental analysis (Carbon) 65.50 to 71.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR