Tryptic soy broth, animal-free, Corning®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
61-411-RO 46-060-LB 46-060-CM 46-060-CI
MDTC61-411-ROEA 194.99 EUR
MDTC61-411-RO MDTC46-060-LB MDTC46-060-CM MDTC46-060-CI
Tryptic soy broth, animal-free, Corning®
Media voor microbiologie Gedehydrateerde media voor microbiologie
This Tryptic Soy Broth (TSB) product is offered for use in sterility testing and validation in the manufacture of a liquid product.
Sterile, ready-to-use solution for the imulaiton of aseptic filling of liquids and sterility testing. Due to the rich nutrient base, TSB may be used in the cultivation of fastidious microorganisms. This growth medium is free of animal-sourced components, making it ideal for testing the sterility of animal-free biopharmaceutical processes.
The use of this medium in place of the product intended for production allows an accurate assessment of sterility throughout the manufacture process.
Certificaten: Complies with current USP requirements.
Order Now
Specificaties
Environmentally Preferable
Shipping temperature Ambient
Opslagtemperatuur Ambient Ambient