Linden grain medium for media fill
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.03790.5000 1.03790.9025
1.03790.5000EA 406 EUR
1.03790.5000 1.03790.9025
Linden grain medium for media fill
Media voor microbiologie Gedehydrateerde media voor microbiologie
Culture medium for media fill simulations of beverage bottling and ice-crusher installations.
  • Ready mixed and easily soluble
  • Granules reduce on-site contamination with media dust
  • Available in 5 kg and cost-saving 25 kg sizes
  • Granulated to minimise toxic and allergenic dust
  • Halal is identical in composition and performance
  • Only available in powder format
When producing low-acid aseptic beverages, you want to be sure that your filling line is not contaminated with spoilage bacteria, yeasts or moulds. Contamination can result in considerable reprocessing, disposal and product recall costs, as well as damage to brand reputation and consumer confidence. Merck granulated ready mixed linden grain medium, a high quality alternative to powdered media, was successfully developed in cooperation with Coca-Cola HBC AG, Zug, Switzerland for media fill simulations.
The unique granulated format of linden grain medium ensures excellent solubility and free flowing properties, means that there is no component separation and clumping, even under warm or humid conditions. It also ensures homogenous distribution of non animal based ingredients and less dust formation, reducing cleaning efforts and limiting the spread of media components that may be hazardous or lead to allergic reactions if inhaled.
Certificaten: Meets highest performance standards in production and QC as described in ISO 11133. Halal certified acc. to JAKIM Standards.