Mannitol salt phenol-red agar for microbiology (According harm. EP/USP/JP) GranuCult®, Millipore®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.05404.0500EA 125 EUR
1.05404.0500 1.05404.5000
Mannitol salt phenol-red agar for microbiology (According harm. EP/USP/JP) GranuCult®, Millipore®
Media voor microbiologie Gedehydrateerde media voor microbiologie
Media is granulated.
  • Granulation preserves homogeneity
  • Easy to weigh out
  • Dissolves faster than powdered media
Granulation preserves homogeneity by capturing the media’s original component distribution in each granule and preventing settling out. Dangers associated with inhalation of fine dust and toxic substances are significantly reduced with granulated media. It is easier to weigh out and dissolves faster than powdered media.