Guzzler® Diaphragm Hand Pump Corregated Hoses
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
06309-10 06309-11
MFLX06309-10EA 64.6 EUR
MFLX06309-10 MFLX06309-11
Guzzler® Diaphragm Hand Pump Corregated Hoses
Slangen Transfer Tubing
Optimize performance of your Guzzler® Diaphragm Hand Pump.
  • Suitable for use with Guzzler® Portable Diaphragm Hand Pumps
These hoses are noncollapsing, wire-reinforced, and constructed of durable vinyl.
Model 06309-10 hose is compatible with Guzzler pumps 07090-09 and -10.
Model 06309-11 hose is compatible with Guzzler pump 07090-11.
Model 06309-12 hose is compatible with Guzzler pump 07090-12.