Polysucrose 400, Corning®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
61-196-RO 61-196-RM
MDTC61-196-ROEA 591 EUR
MDTC61-196-RO MDTC61-196-RM
Polysucrose 400, Corning®
Media voor microbiologie Gedehydrateerde media voor microbiologie
Polysucrose 400 is a synthetic polymer of sucrose and epichlorohydrin used for the density gradient separation of cells, cell membranes and virus cells.
It can also be used to isolate different organelles. This nonionic compound is provided in powder form.