Experimentation kits, hydrophobic and hydrophilic proteins
Leverancier: G-Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BE-404EA 382 EUR
BE-404
Experimentation kits, hydrophobic and hydrophilic proteins
Educatief materiaal Educatief materiaal over biologie Leeractiviteiten voor biochemie
This lab activity is designed to demonstrate the different classes of protein molecules and their classification based on solubility. Students learn fractionation of soluble, insoluble membrane proteins, and cytoskeleton proteins from a tissue sample. The insoluble protein fraction is further fractionated into hydrophilic and hydrophobic membrane proteins. Cell membrane structure and the role of hydrophobic membrane proteins are considered. This lab activity also provides an opportunity to understand characteristics of various classes of detergents and the role of detergents in solubilisation of hydrophobic membrane proteins.
  • Different classes of proteins based on solubility
  • Solubilisation of proteins and role of detergents
  • Characteristics of detergents in aqueous solution
  • Hydrophilic and hydrophobic proteins
  • Membrane proteins and structure of membranes
Also required: Low speed centrifuge for 1.5 to 2 ml tubes, incubator, spectrophotometer and cuvettes or microplate reader and microplate.
Leveringsinformatie: Supplied with components needed for hands-on experimentation for six workstations of 4 to 5 students or 24 to 30 students. Supplied with Teacher’s Guide and separate Student’s Guides.