BloodStor® 100 Freezing Medium
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BloodStor®
STMC07951EA 66 EUR
STMC07951 STMC07939 STMC07938
BloodStor® 100 Freezing Medium
Media voor celbevriezing
Biopreservation reagent for cells and tissues.
  • Sterile, USP vial
  • USP grade/highest-quality components
  • Filled under cGMP
  • Sterility and endotoxin testing
Cryopreserve stem cells and other cells isolated from umbilical cord blood, peripheral blood, bone marrow, and other biologics with BloodStor® 100, an animal component-free and serum-free biopreservation medium. BloodStor® 100 has been sterilised, tested for endotoxin and manufactured under cGMPs with USP-grade components. BloodStor® 100 contains 100% (w/v) DMSO and is available in a variety of formats.