BloodStor® 55-5 Freezing Medium
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BloodStor®
STMC07937EA 790 EUR
STMC07937
BloodStor® 55-5 Freezing Medium
Media voor celbevriezing
Optimised biopreservation reagent for hematopoietic cells and tissues.
  • Ready-to-use
  • Animal component-free
  • Sterile, USP vial
  • cGMP manufactured with USP grade/highest-quality components
  • Sterility, endotoxin, and cell-based release testing
Cryopreserve stem cells and other cells isolated from umbilical cord blood, peripheral blood, bone marrow, and other biologics with BloodStor® 55-5, an animal component-free and ready-to-use medium. cGMP-manufactured with USP-grade components, BloodStor® 55-5 is sterile and tested for cell-based release and endotoxins. BloodStor® 55-5 is pre-formulated with 55% (w/v) DMSO USP, 5% (w/v) Dextran-40 USP, and WFI-quality water. It is available in a variety of formats.