Anti-IgG (H+L) Goat Polyclonal Antibody (iFluor™ 568)
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
STMC100-1083EA 136 EUR
STMC100-1083
Anti-IgG (H+L) Goat Polyclonal Antibody (iFluor™ 568)
Antilichamen
Goat polyclonal antibody against rabbit IgG (H+L).
This goat polyclonal antibody reacts with the heavy (H) chains on rabbit IgG and the light (L) chains common in most rabbit immunoglobulins. Use the polyclonal goat anti-rabbit IgG (H+L) antibody with applications, such as immunohistochemistry (IHC) and immunofluorescence (IF) assays. It has been verified for use with immunocytochemistry (ICC).
Type: Anti-IgG (H+L) Goat Polyclonal Antibody (iFluor™ 568)
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: iFluor™ 568
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit