Human Latent TGF-beta 1 ELISA Kits
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
02018 02019
STMC02018EA 944 EUR
STMC02018 STMC02019
Human Latent TGF-beta 1 ELISA Kits
Testen ELISAs
For detection and measurement of human latent transforming growth factor-beta 1.
The Human Latent Transforming Growth Factor-beta 1 (TGF-β1) Kit is designed for the quantitative detection and measurement of latent human TGF-β1 in biological fluids such as plasma and cell culture supernatants. TGF-β1 is produced primarily by platelets and bone, and in smaller amounts by most nucleated cells and tumours.
TGF-β1 stimulates cells of mesenchymal origin, but inhibits cells of epithelial or neuroectodermal origin. TGF-β1 associates with Latency Associated Protein (LAP) to form a latent complex. This kit detects the LAP entity of the complex. The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the cytokine.
The captured cytokine is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase, which binds the biotinylated antibody. Addition of the chromogenic enzyme substrate 3,3’, 5,5’ tetramethylbenzidine (TMB) results in a coloured product with an intensity directly proportional to the concentration of cytokine in the sample.
The concentration of the cytokine is determined by comparison to a serial dilution of the cytokine standard analysed in parallel.
Opgelet: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').