EasyEights™ EasySep™ Magnet
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EasySep™
STMC18103EA 2838 EUR
STMC18103
EasyEights™ EasySep™ Magnet
Magneten
The EasyEights™ EasySep™ Magnet is designed for cell separation procedures using selected EasySep™ reagents when simultaneously processing multiple samples, up to eight samples on each side, for a total of 16 samples.
  • Magnet for column-free immunomagnetic separation
The EasyEights™ EasySep™ magnet has a large sample range, capable of processing small samples, starting at 0,1×10⁸ cells, to larger samples, up to 8,5×10⁸ cells. The EasyEights™ EasySep™ magnet is designed to hold up to 8 standard 5 ml (12×75 mm) round-bottom tubes on one side, and 8 standard 14 ml (17×95 mm) round-bottom or 15 ml conical tubes on the other side.