EasySep™ Easy 250 Magnet
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EasySep™
STMC100-0821EA 0 EUR
STMC100-0821
EasySep™ Easy 250 Magnet
Magneten
Scale up manual cell isolations from large-volume samples, such as leukopaks and whole blood. Rather than splitting cell suspensions and performing multiple rounds of isolations, the Easy 250 magnet can process up to 225 ml and 1.25×10¹⁰ cells in a single isolation.
  • Magnet for column-free immunomagnetic separation
  • Increase cell isolation throughput by isolating cells from large-volume samples in a single separation
  • Isolate immune cells from samples up to 225 ml in as little as 20 minutes
  • Achieve up to 99% cell purities of viable and functional cells
  • Obtain specific cell types with a wide range of compatible EasySep™ Cell isolation kits
Use the EasySep™ Easy 250 magnet with a standard T-75 cm² cell culture flask and EasySep™ reagents to separate cells from large-volume samples without the use of columns. With its strong, high-gradient internal magnetic field, the magnet isolates cells labeled with magnetic EasySep™ particles, allowing users to simply pipette out the unlabeled cell fraction and obtain cells that are i mmediately ready for downstream applications. Save time and obtain highly purified cells with EasySep™ column-free technology.