EasySep™ Magnet
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EasySep™
STMC18000EA 1368 EUR
STMC18000
EasySep™ Magnet
Magneten
The EasySep™ magnet is designed for cell separation procedures using EasySep™ reagents.
  • Immunomagnetic column-free magnet
  • Designed to hold a standard 12×75 mm (5 ml) polystyrene tube
The EasySep™ magnet generates a high-gradient magnetic field in the interior cavity that is strong enough to separate cells labeled with EasySep™ magnetic particles, without the use of columns.