EasySep™ Easy 50 Magnet
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EasySep™
STMC18002EA 1777 EUR
STMC18002
EasySep™ Easy 50 Magnet
Magneten
The EasySep™ Easy 50 magnet is designed for cell separation procedures using selected EasySep™ reagents when processing a large number of cells (up to 4×10⁹ cells).
  • Immunomagnetic column-free magnet
  • Designed to hold a standard 50 ml conical tube
The EasySep™ Easy 50 magnet generates a high-gradient magnetic field in the interior cavity that is strong enough to separate cells labeled with magnetic EasySep™ particles, without the use of columns.