EasyPlate™ EasySep™ Magnet
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EasyPlate™ EasySep™
STMC18102EA 1271 EUR
STMC18102
EasyPlate™ EasySep™ Magnet
Magneten
The EasyPlate™ EasySep™ Magnet is designed for cell separation procedures using selected EasySep™ reagents when processing a small number of cells (up to 2×10⁷ cells per well).
  • Immunomagnetic column-free magnet
  • Designed to hold a round-bottom, non-tissue culture treated 96-well plate
The EasyPlate™ EasySep™ magnet generates a high-gradient magnetic field that is strong enough to separate cells labeled with EasySep™ D magnetic particles, without the use of columns. The EasyPlate™ EasySep™ magnet is designed to hold a round-bottom, non-tissue culture treated 96-well plate.