Hypoxia Incubator Chamber
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
STMC27310EA 1297 EUR
STMC27310
Hypoxia Incubator Chamber
Testkamers Testkamers voor milieutests
The Hypoxia incubator chamber is for generation of a hypoxic environment for tissue culture. This is a self-contained and sealed chamber that fits inside existing laboratory incubators.
  • All units are molded from high quality polycarbonate and will not break, crack or cloud
  • Integrated stacking feature for storage
  • Guaranteed for 1 year against defects, leaks and breakage
Each unit utilizes a surface-type seal in which all portions of the O-ring are uniformly compressed by a ring clamp for a reliable, air-tight seal. The cylindrical walls and semi-spherical top and bottom minimise gas flow resistance and eliminate dead space during initial purging.
This chamber can use with 84×35 mm plates, 27×60 mm plates, 12×100 mm plates, 12×96-well plates and 18×25 cm² flasks.
Bestelinformatie: Includes with polycarbonate base with O-ring and tubing clamp, polycarbonate lid and tray, ring clamp, O-ring and tubing clamp with tubing.