EasySep™ Human CD14 Positive Selection Kit II
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17858RF 17858 100-0694
STMC17858RFEA 876 EUR
STMC17858RF STMC17858 STMC100-0694
EasySep™ Human CD14 Positive Selection Kit II
Media voor celscheiding
Immunomagnetic positive selection of human CD14+ cells (e.g. monocytes).
  • Fast and easy-to-use
  • Up to 97% purity
  • No columns required
The EasySep™ Human CD14 Positive Selection Kit II is designed to isolate highly purified CD14+ cells from fresh or previously frozen human peripheral blood mononuclear cells (PBMCs). Desired cells are targeted with Tetrameric Antibody Complexes recognising CD14 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube whileMore Product Information