EasySep™ Human EpCAM Positive Selection Kit II
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17846 17846RF
STMC17846EA 668 EUR
STMC17846 STMC17846RF
EasySep™ Human EpCAM Positive Selection Kit II
Media voor celscheiding
Immunomagnetic positive selection of human EpCAM+ cells (e.g. mammary epithelial cells).
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
The EasySep™ Human EpCAM Positive Selection Kit II is designed to isolate EpCAM+ cells from fresh or previously frozen human mammary epithelial cell preparations by immunomagnetic positive selection. Desired cells are targeted with antibody complexes recognising EpCAM and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimise nonspecific binding. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The EpCAM+ fraction contains luminal cells, luminal-restricted, and bipotential progenitor cells.
This product replaces the EasySep™ Human EpCAM Positive Selection Kit (STMC18356).
Opgelet: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).