EasySep™ FITC Positive Selection Kit II
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17682 17682RF
STMC17682EA 504 EUR
STMC17682 STMC17682RF
EasySep™ FITC Positive Selection Kit II
Media voor celscheiding
Immunomagnetic positive selection of any cells labeled with FITC-conjugated antibodies.
  • Fast and easy-to-use
  • No columns required
The EasySep™ FITC Positive Selection Kit II is designed to isolate cells that are labeled with FITC-conjugated antibodies. Desired cells are targeted with antibody complexes recognising FITC and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
This product replaces the EasySep™ FITCMore Product Information