EasySep™ Human Naïve B Cell Isolation Kits
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17254RF 17254
STMC17254RFEA 1232 EUR
STMC17254RF STMC17254
EasySep™ Human Naïve B Cell Isolation Kits
Media voor celscheiding
Immunomagnetic isolation of untouched human naïve B cells.
  • Fast, easy-to-use and column-free
  • Up to 98% purity with high recovery
  • Isolated cells are untouched
The EasySep™ Human Naïve B Cell Isolation Kit is designed to isolate naïve B cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube. This product can be used in place of the EasySep™ Human Naïve B Cell Enrichment Kit (STMC19254) for even faster cell isolations.
Opgelet: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).