RosetteSep™ Human CD3 Depletion Cocktails
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
15621 15661
STMC15621EA 154 EUR
STMC15621 STMC15661
RosetteSep™ Human CD3 Depletion Cocktails
Media voor celscheiding
Immunodensity depletion of CD3+ T cells.
  • Fast and easy-to-use
  • Requires no special equipment or training
  • Untouched, viable cells
  • Can be combined with SepMate™ for consistent, high-throughput sample processing
The RosetteSep™ Human CD3 Depletion Cocktail is designed to deplete CD3+ cells from whole blood. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising CD3 and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as RosetteSep™ DM-L (STMC15705) or Lymphoprep™ (STMC07801), theMore Product Information