RosetteSep™ Human Total Lymphocyte Enrichment Cocktails
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
15263 15223
STMC15263EA 1221 EUR
STMC15263 STMC15223
RosetteSep™ Human Total Lymphocyte Enrichment Cocktails
Media voor celscheiding
Immunodensity isolation of untouched lymphocytes.
  • Fast and easy-to-use
  • Requires no special equipment or training
  • Untouched, viable cells
  • Can be combined with SepMate™ for consistent, high-throughput sample processing
The RosetteSep™ Human Total Lymphocyte Enrichment Cocktail is designed to enrich lymphocytes from whole blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising CD16, CD36, CD66b and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as RosetteSep™ DM-L (STMC15705) or Lymphoprep™ (STMC07801), the unwanted cells pellet along with the RBCs. The purified lymphocytes are present as a highly enriched population at the interface between the plasma and the buoyant density medium.
Opgelet: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).