RosetteSep™ Human Cord Blood Debulking Cocktails
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
15166 15126
STMC15166EA 1133 EUR
STMC15166 STMC15126
RosetteSep™ Human Cord Blood Debulking Cocktails
Media voor celscheiding
Immunodensity depletion of lineage positive cells from cord blood.
  • Fast and easy-to-use
  • Requires no special equipment or training
  • Untouched, viable cells
  • Can be combined with SepMate™ for consistent, high-throughput sample processing
The RosetteSep™ Human Cord Blood Debulking Cocktail is designed to deplete lineage positive cells from fresh cord blood. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising CD2, CD14, CD19, CD66b and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as Lymphoprep™ (STMC07801), the unwanted cells pellet along with the RBCs. The hematopoietic stem and progenitor cells are present as a pre-enriched population at the interface between the plasma and the buoyant density medium.
Opgelet: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).