RosetteSep™ Human Bone Marrow Progenitor Cell Pre-Enrichment Cocktails
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
15067 15027
STMC15067EA 2606 EUR
STMC15067 STMC15027
RosetteSep™ Human Bone Marrow Progenitor Cell Pre-Enrichment Cocktails
Media voor celscheiding
Immunodensity pre-enrichment of untouched stem/progenitor cells from bone marrow.
  • Fast and easy-to-use
  • Requires no special equipment or training
  • Untouched, viable cells
The RosetteSep™ Human Bone Marrow Progenitor Cell Pre-Enrichment Cocktail is designed to pre-enrich progenitor cells by removing lineage-specific cells from whole bone marrow by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising CD3, CD11b, CD14, CD16, CD19, CD56, CD66b and glycophorin A on red blood cells (RBCs). When centrifugedMore Product Information