EasySep™ HLA Whole Blood T Cell Enrichment Kits
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19951HLA 19951HLARF
STMC19951HLAEA 689 EUR
STMC19951HLA STMC19951HLARF
EasySep™ HLA Whole Blood T Cell Enrichment Kits
Media voor celscheiding
Immunomagnetic isolation of untouched human T cells
  • Fast, easy-to-use and column-free
  • Up to 98% purity
  • Untouched, viable cells
  • Compatible with downstream HLA assays
The Complete Processing Kit for whole blood uses EasySep™ HLA T Cell Enrichment Kit to isolate human T cells from HetaSep™-treated whole blood by negative selection. Unwanted cells are targeted for removal with tetrameric antibody complexes recognising non-T cells and dextran-coated magnetic particles. The labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Opgelet: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').