EasySep™ Mouse Streptavidin RapidSpheres™ Isolation Kits
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19860RF 19860
STMC19860RFEA 663 EUR
STMC19860RF STMC19860
EasySep™ Mouse Streptavidin RapidSpheres™ Isolation Kits
Media voor celscheiding
Immunomagnetic isolation of untouched mouse cells
  • Fast and easy-to-use
  • No columns required
  • Untouched, viable cells
The EasySep™ Mouse Streptavidin RapidSpheres™ Isolation Kit is designed for the depletion of single or multiple unwanted cell types labelled with biotinylated antibodies. labelled cells from mouse splenocytes or other tissues are targeted for removal by Streptavidin RapidSpheres™ and separated without the use of columns using an EasySep™ magnet. Desired cells are untouched and poured off into a new tube.
Opgelet: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').