EasySep™ Mouse CD8+ T Cell Isolation Kits
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19853 19853RF
STMC19853EA 867 EUR
STMC19853 STMC19853RF
EasySep™ Mouse CD8+ T Cell Isolation Kits
Media voor celscheiding
Immunomagnetic isolation of untouched mouse CD8+ T cells
  • Fast and easy-to-use
  • Up to 95% purity
  • No columns required
  • Untouched, viable cells
The EasySep™ Mouse CD8+ T Cell Isolation Kit is designed to isolate CD8+ T cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-CD8+ T cells and streptavidin-coated magnetic particles (RapidSpheres™). labelled cells are separated using an EasySep™ magnet without the use of columns. DesiredMore Product Information