EasySep™ Mouse T Cell Isolation Kits
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19851 19851RF
STMC19851EA 855 EUR
STMC19851 STMC19851RF
EasySep™ Mouse T Cell Isolation Kits
Media voor celscheiding
Immunomagnetic isolation of untouched mouse T cells
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
  • Untouched, viable cells
The EasySep™ Mouse T Cell Isolation Kit is designed to isolate T cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-T cells and streptavidin-coated magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.More Product Information