EasySep™ Mouse Naïve CD4+ T Cell Isolation Kits
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19765 19765RF
STMC19765EA 978 EUR
STMC19765 STMC19765RF
EasySep™ Mouse Naïve CD4+ T Cell Isolation Kits
Media voor celscheiding
Immunomagnetic isolation of untouched mouse naive CD4+ T cells
  • Fast, easy-to-use and column-free
  • Up to 95% purity
  • Untouched, viable cells
The EasySep™ Mouse Naïve CD4+ T Cell Isolation Kit is designed to isolate naïve CD4+ T cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-naïve CD4+ T cells , featuring RapidSphere™ technology.
Opgelet: Product Restrictions: (i) Customer shall not make products or any portion of them,More Product Information