EasySep™ Mouse Pan-DC Enrichment Kits
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19763 19763RF
STMC19763EA 980 EUR
STMC19763 STMC19763RF
EasySep™ Mouse Pan-DC Enrichment Kits
Media voor celscheiding
Immunomagnetic isolation of untouched mouse pan-dendritic cells (DCs)
  • Fast, easy-to-use and column-free
  • Up to 76% purity
  • Isolated cells are untouched
The EasySep™ Mouse Pan-DC Enrichment Kit is designed to isolate pan-dendritic cells which isolates cells in just 13 minutes.
Opgelet: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of originMore Product Information