EasySep™ Mouse Neutrophil Enrichment Kits
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19762 19762RF
STMC19762EA 809 EUR
STMC19762 STMC19762RF
EasySep™ Mouse Neutrophil Enrichment Kits
Media voor celscheiding
Immunomagnetic isolation of untouched mouse neutrophils
  • Fast, easy-to-use and column-free
  • Up to 93,7% (blood) and 88,7% (bone marrow) purity
  • Isolated cells are untouched
The EasySep™ Mouse Neutrophil Enrichment Kit is designed to isolate neutrophils from single-cell suspensions of bone marrow or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies that are directed against non-neutrophils. labelled cells are then recognized by Tetrameric Antibody Complexes that are directed against biotin andMore Product Information