EasySep™ Mouse Custom Enrichment Kits
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19709 19709RF
STMC19709EA 0 EUR
STMC19709 STMC19709RF
EasySep™ Mouse Custom Enrichment Kits
Media voor celscheiding
Immunomagnetic isolation of untouched mouse cells
The EasySep™ Mouse Custom Enrichment Kit is designed to isolate any cell type of interest from single-cell suspension of spleen or other tissues by negative selection. STEMCELL Technologies offers a wide selection of antibodies for custom separation and will work with you to isolate a cell type of interest. Please contact our technical support team for more information.
Opgelet: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another productMore Product Information