EasySep™ Direct Human Total Lymphocyte Isolation Kits
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19655 19655RF
STMC19655EA 692 EUR
STMC19655 STMC19655RF
EasySep™ Direct Human Total Lymphocyte Isolation Kits
Media voor celscheiding
Immunomagnetic isolation of total lymphocytes directly from whole blood
  • >99,9% RBC depletion without the need for density gradient centrifugation, sedimentation or lysis
  • Fast, easy-to-use and column-free
  • Up to 96% purity of isolated cells
  • Isolated cells are untouched
The EasySep™ Direct Human Total Lymphocyte Isolation Kit isolates highly purified lymphocytes directly from human whole blood by immunomagnetic negative selection. Isolated cells are untouched and available for direct use in flow cytometry, culture​,​ DNA/RNA extraction or otherMore Product Information