EasySep™ Non-Human Primate CD8+ T Cell Isolation Kits
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19583 19583RF
STMC19583EA 1034 EUR
STMC19583 STMC19583RF
EasySep™ Non-Human Primate CD8+ T Cell Isolation Kits
Media voor celscheiding
Immunomagnetic isolation of untouched non-human primate CD8+ T cells
  • Fast, easy to use, and column-free
  • Up to 85% purity with high recovery
  • Untouched, viable cells
The EasySep™ Non-Human Primate CD8+ T Cell Isolation Kit is designed to isolate CD8+ T cells from fresh peripheral blood mononuclear cells by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labelled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
This kit has been verified for use with rhesus and cynomolgus macaques.
Opgelet: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').