EasySep™ Human Naïve CD8+ T Cell Isolation Kits
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19258RF 19258
STMC19258RFEA 1336 EUR
STMC19258RF STMC19258
EasySep™ Human Naïve CD8+ T Cell Isolation Kits
Media voor celscheiding
Immunomagnetic isolation of untouched human naïve CD8+ T cells
  • Fast, easy-to-use and column-free
  • Up to 97% purity
  • Untouched, viable cells
The EasySep™ Human Naïve CD8+ T Cell Isolation Kit is designed to isolate naïve CD8+ T cells from fresh peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with antibody complexes recognising CD4, CD14, CD16, CD19, CD20, CD36, CD45RO, CD56, CD57, CD94, CD123, CD244, TCRγ/δ, glycophorin A and dextran-coated magnetic particles. The labelled cells are separated using anMore Product Information