EasySep™ Human Pan-DC Pre-Enrichment Kits
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19251 19251RF
STMC19251EA 1070 EUR
STMC19251 STMC19251RF
EasySep™ Human Pan-DC Pre-Enrichment Kits
Media voor celscheiding
Immunomagnetic isolation of untouched human pan dendritic cells (DC)
  • Fast, easy-to-use and column-free
  • Up to 80% purity
  • Untouched, viable cells
The EasySep™ Human Pan-DC Pre-Enrichment Kit is designed to pre-enrich all dendritic cells (DCs) (including myeloid and plasmacytoid DCs) from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising non-pan DCs and dextran-coated magnetic particles. The labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Opgelet: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').