EasySep™ Human Memory CD8+ T Cell Enrichment Kits
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19159RF 19159
STMC19159RFEA 1251 EUR
STMC19159RF STMC19159
EasySep™ Human Memory CD8+ T Cell Enrichment Kits
Media voor celscheiding
Immunomagnetic isolation of untouched human memory CD8+ T cells
  • Fast, easy-to-use and column-free
  • Up to 92% purity
  • Untouched, viable cells
The EasySep™ Human Memory CD8+ T Cell Enrichment Kit is designed to isolate memory CD8+ T cells from peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with antibody complexes recognising CD4, CD14, CD16, CD19, CD20, CD34, CD36, CD45RA, CD56, CD61, CD66b, CD123, TCRγ/δ, glycophorin A and dextran-coated magnetic particles. The labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Opgelet: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').