EasySep™ Mouse CD11c Positive Selection Kit II
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
18780 18781RF 18781 18780RF
STMC18780EA 915 EUR
STMC18780 STMC18781RF STMC18781 STMC18780RF
EasySep™ Mouse CD11c Positive Selection Kit II
Media voor celscheiding
Immunomagnetic positive selection of mouse CD11c+ cells (e.g. dendritic cells (DC)) from mouse splenocytes or other tissues
  • Fast and easy-to-use
  • Up to 95% purity
  • No columns required
  • Isolated cells are not fluorochrome-labelled
The EasySep™ Mouse CD11c Positive Selection Kit II isolates highly purified CD11c+ cells from splenocytes or other tissues by immunomagnetic positive selection. Desired cells are targeted with antibodies and magnetic particles, and isolated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, whileMore Product Information