EasySep™ Human B Cell Enrichment Kits
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19054 19054RF
STMC19054EA 1101 EUR
STMC19054 STMC19054RF
EasySep™ Human B Cell Enrichment Kits
Media voor celscheiding
Immunomagnetic isolation of untouched human B cells.
  • Fast, easy-to-use and column-free
  • Up to 99% purity
  • Untouched, viable cells
The EasySep™ Human B Cell Enrichment Kit is designed to isolate B cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising non-B cells and dextran-coated magnetic particles. The labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a newMore Product Information
Order Now
Specificaties
Environmentally Preferable